Lithium chloride, anhydrous 98+%

Supplier: Thermo Scientific Chemicals
Warning

A10531-0E A10531-22 A10531-36
AAA10531-0EEA 597.64 USD
AAA10531-0E AAA10531-22 AAA10531-36
Lithium chloride, anhydrous 98+%
Lithium chloride

White crystalline powder. Hygroscopic.


Purity: 98+% CAS No.: 7447-41-8 Molecular Formula: LiCl Formula Weight: 42.39 Odor: Odorless Melting Point: 605° Boiling Point: 1325-1360° Merck Reference: 14,5528 MDL No.: MFCD00011078

Formula: LiCl
Boiling Pt: 1325…1360 °C
Melting Pt: 605 °C
Density: 2.068
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 14,5528

Order Now

Specification Test Results

Assay 98.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR