1,2,4-Trichlorobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

A10130-0E A10130-36 A10130-CLL
AAA10130-0EEA 134.03 USD
AAA10130-0E AAA10130-36 AAA10130-CLL
1,2,4-Trichlorobenzene 99%
1,2,4-Trichlorobenzene

MDL: MFCD00000547 Miscible with ether, benzene, CS2


Purity: 99% CAS No.: 120-82-1 Molecular Formula: C6H3Cl3 Formula Weight: 181.45 Flash Point: 110°(230°F) Refractive Index: 1.572 Melting Point: 17° Boiling Point: 213° Solubility: Miscible with ether, benzene, CS2 Merck Reference: 14,9631 UN No.: 2321 MDL No.: MFCD00000547

Formula: C₆H₃Cl₃
Boiling Pt: 213 °C
Melting Pt: 17 °C
Density: 1.454
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000547
CAS Number: 120-82-1
EINECS: 204-428-0
UN: 2321
ADR: "6.1",III
Merck Index: 14,9631

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR