Lithium, AAS Standard Solution, Specpure®, Li 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

088076-AE 088076-AP
AA88076-AEEA 50.61 USD
AA88076-AE AA88076-AP
Lithium, AAS Standard Solution, Specpure®, Li 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Lithium Standards

Molecular Formula: Li2CO3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134051

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR