Iron(II,III) oxide 97%, nanopowder

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Magnetite, Triiron tetraoxide, Iron(II,III) oxide

047141-14 047141-22 047141-36
AA47141-14EA 49.89 USD
AA47141-14 AA47141-22 AA47141-36
Iron(II,III) oxide 97%, nanopowder
Iron (II,III) oxide

50-100nm APS Powder, S. A. 20-50m^2/g.


Purity: 97% CAS No.: 1317-61-9 Molecular Formula: Fe3O4 Formula Weight: 231.54 Physical Form: 50-100nm APS Powder, S. A. 20-50m2/g Density: 5.2 Refractive Index: 3.0 Appearance: Black Odor: Odorless Melting Point: 1538° MDL No.: MFCD00011010

Formula: Fe₃O₄
MW: 231.54 g/mol
Melting Pt: 1560
Density: 5.2
Storage Temperature: Ambient
MDL Number: MFCD00011010
CAS Number: 1317-61-9
EINECS: 215-277-5
Merck Index: 14,4040

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR