Manganese(III) acetate dihydrate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetic acid manganese (III) salt dihydrate, Manganese triacetate dihydrate

047050-04 047050-09
AA47050-04EA 149 USD
AA47050-04 AA47050-09
Manganese(III) acetate dihydrate 97%
Manganese (III) acetate dihydrate

Manganese(III) acetate dihydrate, 97%

Formula: (H₃CCOO)₂Mn·2H₂O
MW: 209.06
Storage Temperature: Ambient
MDL Number: MFCD00150022
CAS Number: 19513-05-4
EINECS: 213-602-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR