Tantalum, gauze woven from wire 100 mesh woven from 0.08mm (0.003in)

Supplier: Thermo Scientific Chemicals

046695-FL
AA46695-FLEA 143 USD
AA46695-FL AA46695-GT
Tantalum, gauze woven from wire 100 mesh woven from 0.08mm (0.003in)
Tantalum

  • Wire Cloth


CAS No.: 7440-25-7 Physical Form: Wire Cloth Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-13505

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR