Zeolite Y, hydrogen

Supplier: Thermo Scientific Chemicals

045870-14 045870-22 045870-36
AA45870-14EA 34 USD
AA45870-14 AA45870-22 AA45870-36
Zeolite Y, hydrogen
Zeolite Y, hydrogen

Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products


CAS No.: 1318-02-1 Molecular Formula: SiO2:Al2O3 Physical Form: Powder, S.A. 780 m2/g, 30:1 mole ratio Appearance: White MDL No.: MFCD00132601


Applications: Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products

Formula: SiO₂:Al₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00132601
CAS Number: 1318-02-1
EINECS: 215-283-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR