Titanium ≥99.99% (metals basis), wire, hard, Diameter 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals
Warning

045485-BU 045485-BY 045485-CC
AA45485-BUEA 56.7 USD
AA45485-BU AA45485-BY AA45485-CC
Titanium ≥99.99% (metals basis), wire, hard, Diameter 1.0 mm (0.04 in)
Titanium

Purity: 99.99% CAS No.: 7440-32-6 Physical Form: ≈3.54g/m Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR