Zinc sulfide ≥99.995% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Zinc (II) sulphide, Zinc (II) sulfide

045454-30 045454-09 045454-18
AA45454-30EA 246 USD
AA45454-30 AA45454-09 AA45454-18
Zinc sulfide ≥99.995% (metals basis)
Zinc sulphide

Purity: 99.995% CAS No.: 1314-98-3 Molecular Formula: SZn Formula Weight: 97.43 Physical Form: 6mm & down pieces Density: 4.10 Appearance: White Odor: Odorless Melting Point: 1700° Merck Reference: 14,10160 MDL No.: MFCD00011301

Formula: SZn
MW: 97.46 g/mol
Melting Pt: 1064
Density: 4.10
Storage Temperature: Ambient
MDL Number: MFCD00011301
CAS Number: 1314-98-3
EINECS: 215-251-3
Merck Index: 14,10160

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR