Copper 65, 65Cu, Plasma Standard Solution, 65Cu 10 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

045276-AD
AA45276-ADEA 144 USD
AA45276-AD
Copper 65, 65Cu, Plasma Standard Solution, 65Cu 10 µg/ml, Thermo Scientific
Standards Cationic Standards Copper Standards

Molecular Formula: Matrix: 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00144006

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR