Mercury, plasma standard solution, Specpure®, Hg 10µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

045259-AE
AA45259-AEEA 110.38 USD
AA45259-AE
Mercury, plasma standard solution, Specpure®, Hg 10µg/ml, Thermo Scientific
Standards Cationic Standards Mercury Standards

Molecular Formula: Hg in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011035

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR