Tin ≥99.9% (metals basis), ingot/button, Diameter ∼36 mm (1.4 in), Thickness 8 mm (0.3 in)

Supplier: Thermo Scientific Chemicals

045110-KF
AA45110-KFEA 56.4 USD
AA45110-KF
Tin ≥99.9% (metals basis), ingot/button, Diameter ∼36 mm (1.4 in), Thickness 8 mm (0.3 in)
Tin

Purity: 99.9% CAS No.: 7440-31-5 Physical Form: ≈50g/piece Appearance: Silver Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR