Nickel ≥99.5% (metals basis), foil, Thickness 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals
Danger

044824-FI 044824-GH 044824-HP
AA44824-FIEA 43.4 USD
AA44824-FI AA44824-GH AA44824-HP
Nickel ≥99.5% (metals basis), foil, Thickness 1.0 mm (0.04 in)
Nickel

Purity: 99.5% CAS No.: 7440-02-0 Physical Form: ≈22.3g/50x50mm Odor: Odorless MDL No.: MFCD00011137

Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR