Graphite ≥99.998% (metals basis), rods, Length 30.5 cm (12 in), Diameter 13cm (5.125 in)

Supplier: Thermo Scientific Chemicals

043649-KF
AA43649-KFEA 1116 USD
AA43649-KF
Graphite ≥99.998% (metals basis), rods, Length 30.5 cm (12 in), Diameter 13cm (5.125 in)
Graphite

Purity: 99.998% Physical Form: ≈7kg/pc MDL No.: MFCD00144065

Formula: C
Melting Pt: 3827 °C
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR