n-Hexane, anhydrous

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

043263-K7 043263-AK 043263-K2
AA43263-K7EA 521.72 USD
AA43263-K7 AA43263-AK AA43263-K2
n-Hexane, anhydrous
n-Hexane

Solvent in Sol-Gel precursor applications


CAS No.: 110-54-3 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -23°(-9°F) Physical Form: Liquid Density: 0.659 Refractive Index: 1.3750 Appearance: Colorless Odor: Weak, characteristic Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 Merck Reference: 14,4694 UN No.: 1208 MDL No.: MFCD00009520


Solvent in Sol-Gel precursor applications

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 69 °C
Melting Pt: –95 °C
Density: 0.659
Flash Pt: –23 °C (–9 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 14,4694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR