Titanium ≥99.995% (metals basis), slug, Puratronic®, Length 3.175 mm (0.125 in), Diameter 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals
Warning

042394-14 042394-06
AA42394-14EA 220 USD
AA42394-14 AA42394-06
Titanium ≥99.995% (metals basis), slug, Puratronic®, Length 3.175 mm (0.125 in), Diameter 3.175 mm (0.125 in)
Titanium

MDL: MFCD00011264


Purity: 99.995% CAS No.: 7440-32-6 Molecular Formula: Ti Physical Form: ≈0.11g/slug Density: 4.5 Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.5
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR