Multi Ion Anion IC Standard Solution in H₂O, Specpure®, Thermo Scientific

Supplier: Thermo Scientific Chemicals

041694-AE
AA41694-AEEA 223.35 USD
AA41694-AE
Multi Ion Anion IC Standard Solution in H₂O, Specpure®, Thermo Scientific
Standards Anionic Standards Multi-Element Anioinic Standards

Physical Form: Liquid
MDL No.: MFCD00144813

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR