Multi Ion Anion IC Standard Solution in H₂O, Specpure®, Thermo Scientific

Supplier: Thermo Scientific Chemicals

041693-AE
AA41693-AEEA 423.94 USD
AA41693-AE
Multi Ion Anion IC Standard Solution in H₂O, Specpure®, Thermo Scientific
Standards Anionic Standards Multi-Element Anioinic Standards

Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00144814

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR