Tin ≥98.8% (metals basis), foil, Thickness 0.05 mm (0.002 in)

Supplier: Thermo Scientific Chemicals

041604-VA 041604-VE 041604-CL
AA41604-VAEA 65 USD
AA41604-VA AA41604-VE AA41604-CL
Tin ≥98.8% (metals basis), foil, Thickness 0.05 mm (0.002 in)
Tin

MDL: MFCD00133862


Purity: 98.8% CAS No.: 7440-31-5 Physical Form: ≈8.2g/15x15cm Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR