Zirconium(IV) oxide ≥99.7% (metals basis excluding Hf) (max. 75ppm Hf)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Zirconium dioxide, Zirconium Oxide

041528-30 041528-A1 041528-18
AA41528-30EA 144.13 USD
AA41528-30 AA41528-A1 AA41528-18
Zirconium(IV) oxide ≥99.7% (metals basis excluding Hf) (max. 75ppm Hf)
Zirconium (IV) oxide

MDL: MFCD00011310


Purity: 99.7% CAS No.: 1314-23-4 Molecular Formula: ZrO2 Formula Weight: 123.22 Physical Form: -325 Mesh Powder Density: 5.89 Melting Point: ca 2700° Boiling Point: ca 5000° Merck Reference: 14,10180 MDL No.: MFCD00011310

Formula: O₂Zr
Boiling Pt: ∼5000 °C
Melting Pt: ∼2700 °C
Density: 5.89
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR