Graphite (natural) ≥99.9995% (metals basis), powder -200 mesh, briquetting grade

Supplier: Thermo Scientific Chemicals
Warning

040795-TB 040795-TC 040795-TD
AA40795-TBEA 226 USD
AA40795-TB AA40795-TC AA40795-TD
Graphite (natural) ≥99.9995% (metals basis), powder -200 mesh, briquetting grade
Graphite

Purity: 99.9995% Physical Form: UCP-1 grade, Ultra "F" purity Appearance: Black Odor: Odorless UN No.: 1325 MDL No.: MFCD00144065

Formula: C
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR