Diyttrium trioxide ≥99.9% (REO, rare earth oxide basis), pellets

Supplier: Thermo Scientific Chemicals

Synonyms: Yttrium oxide, Yttrium (III) oxide

040648-22
AA40648-36EA 1597.53 USD
AA40648-36 AA40648-22
Diyttrium trioxide ≥99.9% (REO, rare earth oxide basis), pellets
Diyttrium trioxide

Sintered pellets suitable for vacuum deposition


Purity: 99.9% CAS No.: 1314-36-9 Molecular Formula: Y2O3 Formula Weight: 225.81 Physical Form: approx. 10mm dia x 5mm thick Density: 5.01 Melting Point: 2410° Merck Reference: 14,10107 MDL No.: MFCD00011473


Sintered pellets suitable for vacuum deposition

Formula: Y₂O₃
Boiling Pt: 4300 °C (1013 hPa)
Melting Pt: 2410 °C
Density: 5.01
Storage Temperature: Ambient
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5
Merck Index: 14,10107

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR