Niobium ≥99.99% (metals basis excluding Ta), powder, Puratronic® -325 mesh (max. 500ppm Ta)

Supplier: Thermo Scientific Chemicals
Danger

040510-09 040510-30 040510-18
AA40510-09EA 68.71 USD
AA40510-09 AA40510-30 AA40510-18
Niobium ≥99.99% (metals basis excluding Ta), powder, Puratronic® -325 mesh (max. 500ppm Ta)
Niobium

MDL: MFCD00011126


Purity: 99.99% CAS No.: 7440-03-1 Physical Form: APS 10 micron Odor: Odorless UN No.: 3089 MDL No.: MFCD00011126

Formula: Nb
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR