Ethyl acetate ≥99.5%, Spectrophotometric Grade for spectrophotometry

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetic acid ethyl ester, Acetic ether, Acetoxyethane, Acetidin, Ethyl acetic ester, Ethyl ethanoate, EtOAc, Vinegar naphtha, Acetic acid ethyl ester

039177-K7 039177-K2 039177-M1
AA39177-K7EA 175 USD
AA39177-K7 AA39177-K2 AA39177-M1
Ethyl acetate ≥99.5%, Spectrophotometric Grade for spectrophotometry
Ethyl acetate

MDL: MFCD00009171 Beilstein Registry No.: 506104 Notes: Meets ACS Spectrophotometric requirements. Filtered through 0.2µ filters.


Purity: 99.5+% CAS No.: 141-78-6 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: -3°(26°F) Physical Form: Liquid Density: 0.902 Refractive Index: 1.3720 Appearance: Colorless Odor: Sweet Melting Point: -84° Boiling Point: 77° Beilstein Reference: 506104 Merck Reference: 14,3757 UN No.: 1173 MDL No.: MFCD00009171

Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1
Melting Pt: -83
Density: 0.902
Flash Pt: –3 °C (26 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 14,3757
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Residual water 0.10%
Impurity content: Chlorinated pesticides 5 ppt
Residue on Evaporation 3 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR