Ground & Waste Water Pollution Standard Solution 3, Specpure®

Supplier: Thermo Scientific Chemicals

036750-AE
AA36750-APEA 303.03 USD
AA36750-AP AA36750-AE
Ground & Waste Water Pollution Standard Solution 3, Specpure®
Standards Environmental Analysis Standards

For US EPA Clean Water Act.


  • 6 Analytes


UN No.: 3264
MDL No.: MFCD02091710

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR