Titanium ≥99.999% (metals basis), plate, Puratronic® 100x100 mm (3.9x3.9 in), Thickness 6 mm (0.2 in)

Supplier: Thermo Scientific Chemicals
Warning

036680-KG
AA36680-KGEA 833 USD
AA36680-KG
Titanium ≥99.999% (metals basis), plate, Puratronic® 100x100 mm (3.9x3.9 in), Thickness 6 mm (0.2 in)
Titanium

MDL: MFCD00011264


Purity: 99.999% CAS No.: 7440-32-6 Physical Form: ≈271g/pc Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR