Scandium, Plasma Standard Solution, Specpure®, Sc 10000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

035755-AP 035755-AD 035755-AE
AA35755-APEA 700 USD
AA35755-AP AA35755-AD AA35755-AE
Scandium, Plasma Standard Solution, Specpure®, Sc 10000 µg/ml, Thermo Scientific
Standards Cationic Standards Scandium Standards

Molecular Formula: Sc2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00016323

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR