Chromium(III) chloride, anhydrous ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chromic chloride, Chromium trichloride

035691-06 035691-22 035691-14
AA35691-06EA 43.4 USD
AA35691-06 AA35691-22 AA35691-14
Chromium(III) chloride, anhydrous ≥99.9% (metals basis)
Chromium (III) chloride

MDL: MFCD00010948


Purity: 99.9% CAS No.: 10025-73-7 Molecular Formula: CrCl3 Formula Weight: 158.35 Physical Form: Flake Density: 2.87 Appearance: Violet Melting Point: ca 1150° Merck Reference: 14,2222 UN No.: 3260 MDL No.: MFCD00010948

Formula: Cl₃Cr
Melting Pt: ∼ 1150 °C
Density: 2.87
Storage Temperature: Ambient
MDL Number: MFCD00010948
CAS Number: 10025-73-7
EINECS: 233-038-3
UN: 3260
ADR: 8,III
Merck Index: 14,2222

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR