Ammonium, Ion chromatography standard solution, Specpure®, NH4+ 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
035560-AE
AA35560-AEEA 136 USD
AA35560-AE
Ammonium, Ion chromatography standard solution, Specpure®, NH4+ 1000µg/ml, Thermo Scientific
Standards Cationic Standards Ammonium Standards
Standard from (NH₄)₂SO₄ in H₂O
Traceable to NIST