Calcium carbonate ≥99.0% ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Marble

033295-36 033295-22
AA33295-36EA 102.47 USD
AA33295-36 AA33295-22
Calcium carbonate ≥99.0% ACS
Calcium carbonate

MDL: MFCD00010906


Purity: 99.0% min CAS No.: 471-34-1 Molecular Formula: CaCO3 Formula Weight: 100.09 Physical Form: Powder Density: 2.930 Refractive Index: 1.6583 Appearance: White Odor: Odorless Melting Point: 800° dec. Merck Reference: 14,1657 MDL No.: MFCD00010906

Formula: CaCO₃
Melting Pt: 800 °C (dec.)
Density: 2.930
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 14,1657

Order Now

Specification Test Results

Insoluble in dilute hydrochloric acid 0.01%
Cl 0.1%
F 0.0015%
SO3 0.01%
NH3 0.003%
Heavy Metals (as Pb) 0.001%
Fe 0.003%
Ba 0.01%
MG 0.02%%
K 0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR