Acetone ≥99.5% semiconductor grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

019392-M1 019392-K2 019392-K7
AA19392-M1EA 140.16 USD
AA19392-M1 AA19392-K2 AA19392-K7
Acetone ≥99.5% semiconductor grade
Acetone

MDL: MFCD00008765 Beilstein Registry No.: 63580 Fieser: 2,13 3,4 6,9


Purity: 99.5% CAS No.: 67-64-1 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -17°(1°F) Physical Form: Liquid Density: 0.791 Refractive Index: 1.3590 Appearance: Colorless Odor: Characteristic Melting Point: -94° Boiling Point: 56° Beilstein Reference: 63580 Merck Reference: 14,66 UN No.: 1090 MDL No.: MFCD00008765

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4
Density: 0.791
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 14,66
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10
Evaporation residue 5 ppm
Acidity 0.3 µeq/g
Alkalinity 0.5 µeq/g
H2O 0.5%
Solubility in H2O Pass test
Cl 0.2 ppm
PO3 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR