Sodium hydrogen carbonate 99.7-100.3% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium bicarbonate

014707-36 014707-A9 014707-A3
AA14707-36EA 46.66 USD
AA14707-36 AA14707-A9 AA14707-A3
Sodium hydrogen carbonate 99.7-100.3% ACS
Sodium hydrogen carbonate

As a source of CO2, in making sodium salts, baking powder, cleaning compounds


Purity: 99.7-100.3% CAS No.: 144-55-8 Molecular Formula: NaHCO3 Formula Weight: 84.01 Physical Form: Granular Density: 2.159 Refractive Index: 1.500 Odor: Odorless pH: 8,1-8,4 Solubility: Soluble in water. Insoluble in alcohol Beilstein Reference: 4153970 Merck Reference: 14,8583 MDL No.: MFCD00003528


As a source of CO2, in making sodium salts, baking powder, cleaning compounds

Formula: NaHCO₃
Melting Pt: 270 °C
Density: 2.159
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 14,8583
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble matter 0.015%
Cl 0.3%
PO8 0.001%
SO3 0.003%
NH3 5 ppm max.
Ca 0.02%
MG 0.005%
Heavy metals (as Pb) 5 ppm
Fe 0.001%
K 0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR