Potassium bromide, ultra dry ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

014700-14 014700-22 014700-36
AA14700-14EA 91.5 USD
AA14700-14 AA14700-22 AA14700-36
Potassium bromide, ultra dry ≥99.9% (metals basis)
Potassium bromide

MDL: MFCD00011358 Fieser: 21,358


Purity: 99.9% CAS No.: 7758-02-3 Molecular Formula: KBr Formula Weight: 119.01 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 2.75 Refractive Index: 1.559 Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 730° Boiling Point: 1435° Merck Reference: 14,7618 MDL No.: MFCD00011358

Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Density: 2.75
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 14,7618

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR