Telluric acid dihydrate 99

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hydrogen tellurate

014197-18 014197-09
AA14197-18EA 221.27 USD
AA14197-18 AA14197-09
Telluric acid dihydrate 99
Telluric acid dihydrate

Purity: 99+% CAS No.: 13451-14-4 Molecular Formula: H2O4Te.2H2O Formula Weight: 229.64 (193.61anhy) Physical Form: Crystalline Density: 3.158 Sensitivity: Hygroscopic Appearance: White Melting Point: 136° Boiling Point: 160° dec. UN No.: 3284 MDL No.: MFCD00150387

Formula: H₂O₄Te.2H₂0
MW: 229.64 g/mol (193.61 anhy)
Boiling Pt: 160 °C (dec.)
Melting Pt: 136 °C
Density: 3.158
Storage Temperature: Ambient
MDL Number: MFCD00150387
CAS Number: 13451-14-4
EINECS: 232-267-6
UN: 3284
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR