Palladium 5% on activated carbon paste (50% water) A503023-5

Supplier: Thermo Scientific Chemicals

011694-14 011694-22 011694-06
AA11694-14EA 414 USD
AA11694-14 AA11694-22 AA11694-06
Palladium 5% on activated carbon paste (50% water) A503023-5
Palladium

Applications: Debenzylation
Sold on a dry weight basis. Unit weight excludes water weight


CAS No.: 7440-05-3 Appearance: Black Odor: Odorless MDL No.: MFCD03457879


Applications: Debenzylation

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR