Aluminium 99+% (metals basis), wire, annealed, Diameter 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminum

011064-H7 011064-G6 011064-H2
AA11064-H7EA 327 USD
AA11064-H7 AA11064-G6 AA11064-H2
Aluminium 99+% (metals basis), wire, annealed, Diameter 1.0 mm (0.04 in)
Aluminium

MDL: MFCD00134029


Purity: 99+% CAS No.: 7429-90-5 Physical Form: ≈2.1g/m, Temper: soft Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR