Zinc selenide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc (II) selenide

010999-30 010999-18 010999-09
AA10999-30EA 1106 USD
AA10999-30 AA10999-18 AA10999-09
Zinc selenide ≥99.999% (metals basis)
Zinc selenide

MDL: MFCD00011296


Purity: 99.999% CAS No.: 1315-09-9 Molecular Formula: ZnSe Formula Weight: 144.33 Physical Form: Powder Density: 5.42 Melting Point: 1100° Merck Reference: 14,10156 UN No.: 3283 MDL No.: MFCD00011296

Formula: SeZn
MW: 144.35 g/mol
Melting Pt: 1793
Density: 5.42
Storage Temperature: Ambient
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1 III
Merck Index: 14,10156

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR