Sodium sulfate ≥99.9955% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals

Synonyms: Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt

010874-14 010874-22
AA10874-14EA 228 USD
AA10874-14 AA10874-22
Sodium sulfate ≥99.9955% (metals basis), Puratronic®
Sodium sulphate

MDL: MFCD00003504 Fieser: 1,1103


Purity: 99.9955% CAS No.: 7757-82-6 Molecular Formula: Na2SO4 Formula Weight: 142.04 Physical Form: Powder Density: 2.680 Refractive Index: 1.484 Sensitivity: Hygroscopic Odor: Odorless Melting Point: 884° Merck Reference: 14,8680 MDL No.: MFCD00003504

Formula: Na₂SO₄
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 884 °C
Density: 2.680
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 14,8680
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR