Silver nitrate ≥99.9995% (metals basis), Premion®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

010858-14 010858-06 010858-22
AA10858-14EA 256 USD
AA10858-14 AA10858-06 AA10858-22
Silver nitrate ≥99.9995% (metals basis), Premion®
Silver nitrate

MDL: MFCD00003414


Purity: 99.9995% CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Crystalline Density: 4.352 Appearance: White Odor: Odorless Melting Point: 212° Merck Reference: 14,8518 UN No.: 1479 MDL No.: MFCD00003414

Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
Density: 4.352
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1479
ADR: 5.1 I
REACH: 01-2119513705-43
Merck Index: 14,8518
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR