Niobium(V) oxide ≥99.9985% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diniobium pentaoxide, Niobium pentoxide

010822-18 010822-30 010822-A1
AA10822-18EA 191.82 USD
AA10822-18 AA10822-30 AA10822-A1
Niobium(V) oxide ≥99.9985% (metals basis), Puratronic®
Niobium (V) oxide

Powder. Purity based on metal contaminates only.


Purity: 99.9985% CAS No.: 1313-96-8 Molecular Formula: Nb2O5 Formula Weight: 265.81 Physical Form: Powder Density: 4.6 Melting Point: 1520° Merck Reference: 14,6559 MDL No.: MFCD00011128

Formula: Nb₂O₅
Melting Pt: 1520 °C
Density: 4.6
Storage Temperature: Ambient
MDL Number: MFCD00011128
CAS Number: 1313-96-8
EINECS: 215-213-6
Merck Index: 14,6559

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR