Zinc ≥99.9999% (metals basis), shot, Puratronic® 1-6 mm (0.04-0.24 in)

Supplier: Thermo Scientific Chemicals

010760-A1 010760-18 010760-30
AA10760-A1EA 2831 USD
AA10760-A1 AA10760-18 AA10760-30
Zinc ≥99.9999% (metals basis), shot, Puratronic® 1-6 mm (0.04-0.24 in)
Zinc

MDL: MFCD00011291


Purity: 99.9999% CAS No.: 7440-66-6 Odor: Odorless MDL No.: MFCD00011291

Formula: Zn
Boiling Pt: 908 °C (1013 hPa)
Melting Pt: 420 °C
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now

Specification Test Results

Assay (by Metals Basis) 99.9999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR