Titanium ≥99.4%, powder -100 mesh

Supplier: Thermo Scientific Chemicals
Warning

010383-14 010383-22 010383-36
AA10383-14EA 42.7 USD
AA10383-14 AA10383-22 AA10383-36
Titanium ≥99.4%, powder -100 mesh
Titanium

MDL: MFCD00011264


Purity: 99.4% CAS No.: 7440-32-6 Appearance: Grey Odor: Odorless UN No.: 2546 MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3277 °C (1013 hPa)
Melting Pt: 1677 °C
Density: 4,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
UN: 2546
ADR: 4.2,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR