Graphite ≥99% (metals basis), rods, Length 61 cm (24 in), Diameter 1.27cm (0.5 in)

Supplier: Thermo Scientific Chemicals

010134-KJ 010134-KG
AA10134-KJEA 152.28 USD
AA10134-KJ AA10134-KG
Graphite ≥99% (metals basis), rods, Length 61 cm (24 in), Diameter 1.27cm (0.5 in)
Graphite

MDL: MFCD00144065


Purity: 99% Physical Form: ≈130g/pc MDL No.: MFCD00144065

Formula: C
Melting Pt: 3827 °C
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR