Gold ≥99.9% (metals basis), wire, Diameter 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

000725-G1 000725-G5
AA00725-G1EA 276 USD
AA00725-G1 AA00725-G5
Gold ≥99.9% (metals basis), wire, Diameter 0.25 mm (0.01 in)
Gold

MDL: MFCD00003436


Purity: 99.9% Physical Form: ≈0.95g/m Odor: Odorless MDL No.: MFCD00003436

Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR