Palladium, black ≥99.9%

Supplier: Thermo Scientific Chemicals
Danger

000659-06 000659-03
AA00659-06EA 1557 USD
AA00659-06 AA00659-03
Palladium, black ≥99.9%
Palladium

Purity: 99.9% CAS No.: 7440-05-3 Physical Form: S.A. typically 20m2/g Odor: Odorless UN No.: 3089 MDL No.: MFCD00011167

Formula: Pd
MW: 106.420 g/mol
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 3089
ADR: 4.1 II

Order Now

Specification Test Results

Form S.A. typi ~ lly 20m2/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR