Anti-BCL10 Antibody [clone: 151]
Supplier: Diagnostic Biosystems
MOB442-05 MOB442
95024-248EA 336.44 USD
95024-248 99991-488
Anti-BCL10 Antibody [clone: 151]
Antibodies
Monoclonal anti-bcl-10 reacts specifically with human bcl-10. The epitope recognized by the antibody resides within amino acids 122-168 of human bcl-10 molecule. Bcl-10, also designated as CIPER, mE10, cE10, CARMEN, and CLAP, is an N-terminal CARD (caspase recruitment domain) containing protein. It is a cellular homologue of the equine herpesvirus-2 protein E-10 (vCLAP). Bcl-10 was implicated in the regulation of apoptosis by interacting with caspase 9, enhancing procaspase 9 processing, and triggering its activation when overexpressed in the cell. Bcl-10 cellular overexpression induces JNK,More Product Information