Anti-IgG Polyclonal Antibody

Supplier: Diagnostic Biosystems

RP023 RP023-05 PDR018
99991-242EA 414.18 USD
99991-242 95025-060 99991-076
Anti-IgG Polyclonal Antibody
Antibodies
This antibody reacts with γ-chains of human IgG. Nonspecific antibodies have been removed by solid-phase adsorption.

Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR