Anti-CD3 (T Cell) Rabbit Polyclonal Antibody

Supplier: Diagnostic Biosystems

PDR002
99991-044EA 429.41 USD
99991-044
Anti-CD3 (T Cell) Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: CD3 (T Cell)
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR