Anti-IgL Mouse Antibody [clone: N10/2]

Supplier: Diagnostic Biosystems

MOB077 MOB077-05 PDM056
99990-260EA 356.3 USD
99990-260 95024-632 99990-902
Anti-IgL Mouse Antibody [clone: N10/2]
Antibodies
This antibody does not show any cross-reactivity with kappa light chain. It stains B cell follicles in human lymphoid tissues. The mantle zone shows a mosaic pattern of labeling while the germinal centers show a coarse meshwork pattern of staining.

Type:
Antigen: IgL
Clonality: Monoclonal
Clone: N10/2
Conjugation:
Epitope: Light Chain
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR