Anti-CD68 Mouse Monoclonal Antibody [clone: PG-M1]

Supplier: Diagnostic Biosystems

MOB094 MOB094-05 PDM065
99990-288EA 948.36 USD
99990-288 95024-648 99990-916
Anti-CD68 Mouse Monoclonal Antibody [clone: PG-M1]
Antibodies
This antibody reacts with a glycoprotein of 110 kDa, expressed as intracytoplasmic molecule. It stains macrophages in a wide variety of tissues. Myeloid precursor cells and peripheral granulocytes are negative. The PG-M1 differs from EMB11 (CD68) because of its non-reactivity with granulocytes and their precursor cells.

Type:
Antigen: CD68
Clonality: Monoclonal
Clone: PG-M1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR